EMAIL THIS PAGE TO A FRIEND
33031-U Supelco

BSA+TMCS+TMSI

3:2:3, pkg of 1 × 25 mL

Synonym: Chlorotrimethylsilane, N,O-Bis(trimethylsilyl)acetamide, TSIM, Trimethylchlorosilane, Trimethylsilyl chloride, Trimethylsilylimidazole

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Derivatization Reagents, Derivatization Reagents GC, Reagents for Silylation,
grade   for GC/MS (SIM)
description   3:2:3 (SYLON BTZ)
reaction suitability   reagent type: derivatization reagent
reaction type: Acylations
  reagent type: derivatization reagent
reaction type: Silylations
packaging   pkg of 1 × 25 mL

Description

General description

Reagent mixture consisting of BSA+TMCS+TMSI when heated with pyridine and steroid at 60°C for 12 hours, converts 11- and the tertiary 17-hydroxyl groups to trimethylsilyl ethers.

Application

It was used during derivatization of the sample for multi-mycotoxin analysis using gas chromatography (GC) coupled to triple quadrupole instrument (QqQ). It was also used as derivatizing reagent in the determination of trichothecenes, patulin and zearalenone using GC-MS/MS.

Other Notes

Reagent for trifluoroacetate, trimethylsilyl (TMS).

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 2924 8(3) / PGII
WGK Germany 
3
Flash Point(F) 
28.4 °F
Flash Point(C) 
-2 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Silylating Reagents for GC-MS Compatible Derivatives of NSAIDs

Human and veterinary pharmaceuticals are a group of “emerging” contaminants,1, 2 some of which are produced in increasingly large volumes every year. The amounts produced are reaching quantities simi...
Luis-Alberto Martin
AnalytiX Volume 10 Article 2
Keywords: Anti-inflammatory agents, Derivatizations, Environmental, Gas chromatography, Gas chromatography mass spectrometry, Mass spectrometry, Metabolites, Pesticides, Pharmaceutical, Silylations, Veterinary

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

85432 Silylating mixture II according to Horning, for GC derivatization, LiChropur

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?