• USA Home
  • 33100AST - Astec® (S,S) P-CAP Chiral HPLC Guard Column

33100AST Supelco

Astec® (S,S) P-CAP Chiral HPLC Guard Column

Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm

  •  eCl@ss 32110501



Related Categories Analytical/Chromatography, Astec P-CAP, Astec P-CAP and P-CAP-DP Chiral HPLC Columns (Polycyclic Amine Polymers), Chiral HPLC Columns, Chiral HPLC Guard Columns and Accessories,
description   HPLC guard column
packaging   pkg of 1 ea
application(s)   HPLC: suitable
L × I.D.   2 cm × 4 mm
matrix active group   poly(trans-1,2-cyclohexanediyl-bis-acrylamide) phase
particle size   5 μm
pore size   200 Å
mode of chromatography   chiral


Legal Information

Astec is a registered trademark of Sigma-Aldrich Co. LLC

P-CAP is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Milli-Q® Water Purification Solutions
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.


Certificate of Analysis

Protocols & Articles


Chiral HPLC for Chemists: Ultimate Solvent Choice with High Capacity Using Astec’s P-CAP™ Series

The polyamide chiral stationary phase (CSP) features a thin, ordered polymer layer, chemically bonded to 5 μm or 3.5 μm spherical silica, using a patented radical polymerization. This results in two ...
Denise Wallworth
Reporter EU Volume 28
Keywords: Applications, Buffers, High performance liquid chromatography, Mass spectrometry, Phase transitions, Polymerization reactions, Purification, Radical polymerization, Separation, Solvents, Supercritical fluid chromatography, Type

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?