• USA Home
  • 33102-U - Vials, crimp top, serum bottle
EMAIL THIS PAGE TO A FRIEND
33102-U Supelco
Vials, crimp top, serum bottle
volume 5 mL, O.D. × H 25 mm × 40 mm, clear glass bottle, pkg of 36 ea
Purchase
Properties
Related Categories Analytical/Chromatography, Autosampler Vials, Storage Kits and Accessories, Serum Bottles, 5 mL to 100 mL, Supelco glass serum bottles, Vials More...
material   clear glass bottle
feature   autoclavable
  closure type crimp top vial
packaging   pkg of 36 ea
O.D. × H   25 mm × 40 mm
volume   5 mL
fitting   opening 20 mm
Description
General description
These vials are constructed of clear, borosilicate glass; closure not included.
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?