EMAIL THIS PAGE TO A FRIEND
33180-U Supelco

Hand Decapper

for use with 8mm crimp seals, pkg of 1 ea

  •  NACRES NB.11

Purchase

Properties

Related Categories Analytical/Chromatography, Crimpers and Decappers, Hand decapper, Vial Accessories, Vials More...
description   Hand decapper
packaging   pkg of 1 ea
compatibility   for use with 8mm crimp seals
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

33272-U Crimpers, Hand crimper, for use with 8mm seals, pkg of 1 ea

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?