EMAIL THIS PAGE TO A FRIEND
40076 Supelco

Anthracene solution

certified reference material, 1000 μg/mL in acetone

  • CAS Number 120-12-7

  • Empirical Formula (Hill Notation) C14H10

  • Molecular Weight 178.23

  •  Beilstein/REAXYS Number 1905429

  •  MDL number MFCD00001240

  •  PubChem Substance ID 24864949

Purchase

Properties

Related Categories A-B, Additional Standards, All Polycyclic Aromatic Hydrocarbon (PAH) Standards, Alpha Sort, Analytical Standards,
Quality Level   100
grade   certified reference material
  TraceCERT®
packaging   ampule of 1 mL
concentration   1000 μg/mL in acetone
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Environmental
format   single component solution
storage temp.   2-8°C
SMILES string   c1ccc2cc3ccccc3cc2c1
InChI   1S/C14H10/c1-2-6-12-10-14-8-4-3-7-13(14)9-11(12)5-1/h1-10H
InChI key   MWPLVEDNUUSJAV-UHFFFAOYSA-N
Gene Information   human ... CYP1A2(1544)

Description

Application

Anthracene is a solid polycyclic aromatic hydrocarbon, which can undergo sulfonation in glacial acetic acid to obtain a mixture of 1-anthracenesulfonic acid and 2-anthracenesulfonic acid.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
Repeated exposure may cause skin dryness or cracking.
RIDADR 
UN1090 - class 3 - PG 2 - Acetone
WGK Germany 
1
Flash Point(F) 
1.4 °F
Flash Point(C) 
-17 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?