EMAIL THIS PAGE TO A FRIEND
40101 Supelco

β−BHC solution

certified reference material, 1000 μg/mL in acetone

  • CAS Number 319-85-7

  • Empirical Formula (Hill Notation) C6H6Cl6

  • Molecular Weight 290.83

Purchase

Properties

Related Categories A-B, Additional Standards, Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography,
Quality Level   100
grade   certified reference material
  TraceCERT®
CofA   certificate of analysis is enclosed in each package.
packaging   ampule of 1 mL
concentration   1000 μg/mL in acetone
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
format   single component solution
storage temp.   2-8°C
InChI   1S/C6H6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-6H
InChI key   JLYXXMFPNIAWKQ-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
Repeated exposure may cause skin dryness or cracking.
RIDADR 
UN1090 - class 3 - PG 2 - Acetone, solution
WGK Germany 
2
Flash Point(F) 
1.4 °F
Flash Point(C) 
-17 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

GC Analysis of Multiple Pesticides on SLB®-5ms after SPME using 60 μm Carbowax® Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Multiple Pesticides on SLB®-5ms after SPME using 60 μm Carbowax® Fiber
Keywords: Agriculture, Chromatography, Gas chromatography, Metabolites, Pesticides, Purification, Solid phase microextractions

GC Analysis of Organochlorine Pesticides in Water on SPB®-5 after SPME using 100 μm PDMS Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Organochlorine Pesticides in Water on SPB®-5 after SPME using 100 μm PDMS Fiber
Keywords: Chromatography, Electron capture detector, Environmental, Gas chromatography, Metabolites, Pesticides, Purification, Size-exclusion chromatography, Solid phase microextractions

GC Analysis of a 20-Component Organochlorine Pesticide Mix in Water on a 5% Phenyl Column after SPE using Supelclean™ ENVI™-8

From our library of Articles, Sigma-Aldrich presents GC Analysis of a 20-Component Organochlorine Pesticide Mix in Water on a 5% Phenyl Column after SPE using Supelclean™ ENVI™-8
Keywords: Chromatography, Electron capture detector, Gas chromatography, Purification, Sample preparations, Size-exclusion chromatography, Solid phase extractions

US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on Equity®-1701 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on Equity®-1701 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Keywords: Chromatography, Electron capture detector, Gas chromatography, Pesticides, Purification, Size-exclusion chromatography

US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on SLB®-5ms (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on SLB®-5ms (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Keywords: Chromatography, Electron capture detector, Gas chromatography, Pesticides, Purification, Size-exclusion chromatography

US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on SLB®-5ms
Keywords: Chromatography, Electron capture detector, Gas chromatography, Pesticides, Purification

US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on SPB®-608

From our library of Articles, Sigma-Aldrich presents US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on SPB®-608
Keywords: Chromatography, Electron capture detector, Gas chromatography, Pesticides, Purification

US EPA Method 625: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 625: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8081: GC Analysis of Organochlorine Pesticides on Equity®-5

From our library of Articles, Sigma-Aldrich presents US EPA Method 8081: GC Analysis of Organochlorine Pesticides on Equity®-5
Keywords: Chromatography, Electron capture detector, Environmental, Gas chromatography, Pesticides, Purification, Size-exclusion chromatography

US EPA Method TO-10/IP-8 and ASTM® D4861: GC Analysis of Pesticides on SPB®-608 after Collection/Desorption using ORBO™-1000

From our library of Articles, Sigma-Aldrich presents US EPA Method TO-10/IP-8 and ASTM® D4861: GC Analysis of Pesticides on SPB®-608 after Collection/Desorption using ORBO™-1000
Keywords: Air Monitoring, Chromatography, Electron capture detector, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?