• USA Home
  • 40171 - 1,2-Dibromoethane solution

EMAIL THIS PAGE TO A FRIEND
40171 Supelco

1,2-Dibromoethane solution

certified reference material, 5000 μg/mL in methanol

Synonym: Ethylene dibromide

  • CAS Number 106-93-4

  • Empirical Formula (Hill Notation) C2H4Br2

  • Molecular Weight 187.86

  •  MDL number MFCD00000233

  •  PubChem Substance ID 24864873

Purchase

Properties

Quality Level   100
grade   certified reference material
  TraceCERT®
packaging   ampule of 1 mL
concentration   5000 μg/mL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
Environmental
format   single component solution
storage temp.   2-8°C
SMILES string   BrCCBr
InChI   1S/C2H4Br2/c3-1-2-4/h1-2H2
InChI key   PAAZPARNPHGIKF-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
3
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

GC Analysis of Volatiles on SPB®-624, Direct Injection

From our library of Articles, Sigma-Aldrich presents GC Analysis of Volatiles on SPB®-624, Direct Injection
Keywords: Chromatography, Gas chromatography, Purification, Size-exclusion chromatography

GC Analysis of Volatiles on VOCOL® after SPME using 75 μm Carboxen/PDMS Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Volatiles on VOCOL® after SPME using 75 μm Carboxen/PDMS Fiber
Keywords: Chromatography, Gas chromatography, Mass spectrometry, Purification, Size-exclusion chromatography, Solid phase microextractions

US EPA Method 502.2: GC Analysis of Volatiles on VOCOL® (105 m x 0.53 mm I.D.) after Purge & Trap using "K" Trap, ELCD Signal

From our library of Articles, Sigma-Aldrich presents US EPA Method 502.2: GC Analysis of Volatiles on VOCOL® (105 m x 0.53 mm I.D.) after Purge & Trap using "K" Trap, ELCD Signal
Keywords: Chromatography, Gas chromatography, Hall electrolytic conductivity detector, Photo-ionization detector, Purge and trap, Sample preparations

US EPA Method 502.2: GC Analysis of Volatiles on VOCOL® (105 m x 0.53 mm I.D.) after Purge & Trap using "K" Trap, PID Signal

From our library of Articles, Sigma-Aldrich presents US EPA Method 502.2: GC Analysis of Volatiles on VOCOL® (105 m x 0.53 mm I.D.) after Purge & Trap using "K" Trap, PID Signal
Keywords: Chromatography, Gas chromatography, Hall electrolytic conductivity detector, Photo-ionization detector, Purification

US EPA Method 524.2: GC Analysis of Volatiles on VOCOL® (105 m x 0.53 mm I.D.) after Purge & Trap using "K" Trap

From our library of Articles, Sigma-Aldrich presents US EPA Method 524.2: GC Analysis of Volatiles on VOCOL® (105 m x 0.53 mm I.D.) after Purge & Trap using "K" Trap
Keywords: Chromatography, Gas chromatography, Purification, Size-exclusion chromatography

US EPA Method 8260: GC Analysis of Volatiles on VOCOL® after Purge & Trap using "K" Trap, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8260: GC Analysis of Volatiles on VOCOL® after Purge & Trap using "K" Trap, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification, Solid phase microextractions

US EPA Method 8260: GC Analysis of Volatiles on VOCOL® after Purge & Trap using "K" Trap

From our library of Articles, Sigma-Aldrich presents US EPA Method 8260: GC Analysis of Volatiles on VOCOL® after Purge & Trap using "K" Trap
Keywords: Chromatography, Gas chromatography, Purification, Solid phase microextractions

US EPA Method OLM04.2 VOA: GC Analysis of Volatiles on SPB®-624 after Purge & Trap using "K" Trap, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method OLM04.2 VOA: GC Analysis of Volatiles on SPB®-624 after Purge & Trap using "K" Trap, Fast GC Analysis
Keywords: Chromatography, Environmental, Gas chromatography, Purification, Solid phase microextractions

US EPA Method OLM04.2 VOA: GC Analysis of Volatiles on VOCOL® after Purge & Trap using "K" Trap, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method OLM04.2 VOA: GC Analysis of Volatiles on VOCOL® after Purge & Trap using "K" Trap, Fast GC Analysis
Keywords: Chromatography, Environmental, Gas chromatography, Purification, Solid phase microextractions

US EPA Method TO-17: GC Analysis of Volatiles on Equity®-1 after Collection/Desorption using Carbotrap 300 Tube

From our library of Articles, Sigma-Aldrich presents US EPA Method TO-17: GC Analysis of Volatiles on Equity®-1 after Collection/Desorption using Carbotrap 300 Tube
Keywords: Chromatography, Gas chromatography

US EPA Method TO-17: GC Analysis of Volatiles on VOCOL® after Collection/Desorption using Air Toxics Tube

From our library of Articles, Sigma-Aldrich presents US EPA Method TO-17: GC Analysis of Volatiles on VOCOL® after Collection/Desorption using Air Toxics Tube
Keywords: Air Monitoring, Chromatography, Flame ionization detector, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?