• USA Home
  • 442219 - 1,2,4,5-Tetrachlorobenzene

EMAIL THIS PAGE TO A FRIEND
442219 Supelco

1,2,4,5-Tetrachlorobenzene

analytical standard

  • CAS Number 95-94-3

  • Empirical Formula (Hill Notation) C6H2Cl4

  • Molecular Weight 215.89

  •  Beilstein Registry Number 1618315

  •  EC Number 202-466-2

  •  MDL number MFCD00000549

  •  PubChem Substance ID 24867741

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, Chromatography,
grade   analytical standard
InChI Key   JHBKHLUZVFWLAG-UHFFFAOYSA-N
form   neat
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
bp   240-246 °C(lit.)
mp   138-140 °C (lit.)
Featured Industry   Environmental
format   neat
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

General description

1,2,4,5-Tetrachlorobenzene can undergo palladium catalyzed hydrodehalogenation to yield benzene, and the progress of the reaction can be determined chemical kinetics.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
DB9450000
Flash Point(F) 
320 °F
Flash Point(C) 
160 °C
Protocols & Articles

Articles

US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on Equity®-5 (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on Equity®-5 (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Mass selective detector, Purification

US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?