• USA Home
  • 442257 - 1-Chloro-4-fluorobenzene

EMAIL THIS PAGE TO A FRIEND
442257 Supelco

1-Chloro-4-fluorobenzene

analytical standard

Purchase

Properties

grade   analytical standard
InChI Key   RJCGZNCCVKIBHO-UHFFFAOYSA-N
packaging   ampule of 1000 mg
refractive index   n20/D 1.495(lit.)
bp   129-130 °C(lit.)
mp   −27-−26 °C (lit.)
density   1.226 g/mL at 25 °C(lit.)
Featured Industry   Environmental
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
84.2 °F
Flash Point(C) 
29 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?