• USA Home
  • 442259-U - 1-Chloronaphthalene

EMAIL THIS PAGE TO A FRIEND
442259-U Supelco

1-Chloronaphthalene

technical grade

  • CAS Number 90-13-1

  • Empirical Formula (Hill Notation) C10H7Cl

  • Molecular Weight 162.62

  •  Beilstein/REAXYS Number 970836

  •  EC Number 201-967-3

  •  MDL number MFCD00003874

  •  PubChem Substance ID 329756370

Purchase

Properties

Related Categories A-C, Analytical Standards, Analytical/Chromatography, Organic Pollutant Standards, Volatile & Semivolatile Standards More...
grade   technical grade
InChI Key   JTPNRXUCIXHOKM-UHFFFAOYSA-N
form   neat
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.632 (lit.)
bp   111-113 °C/5 mmHg(lit.)
mp   −20 °C (lit.)
density   1.194 g/mL at 25 °C (lit.)
Featured Industry   Environmental
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Components

Remaining % is 2-chloronaphthalene.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
3
RTECS 
QJ2100000
Flash Point(F) 
249.8 °F
Flash Point(C) 
121 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?