EMAIL THIS PAGE TO A FRIEND
442267-U Supelco

1-Heptene

analytical standard

  • CAS Number 592-76-7

  • Linear Formula CH3(CH2)4CH=CH2

  • Molecular Weight 98.19

  •  Beilstein/REAXYS Number 1098332

  •  EC Number 209-767-8

  •  MDL number MFCD00009531

  •  PubChem Substance ID 329756371

  •  NACRES NA.24

Purchase

Properties

Quality Level   100
grade   analytical standard
vapor pressure   101 mmHg ( 37.7 °C)
form   neat
autoignition temp.   505 °F
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.400 (lit.)
bp   94 °C (lit.)
mp   −119 °C (lit.)
density   0.697 g/mL at 25 °C (lit.)
Featured Industry   Petroleum
format   neat
SMILES string   CCCCCC=C
InChI   1S/C7H14/c1-3-5-7-6-4-2/h3H,1,4-7H2,2H3
InChI key   ZGEGCLOFRBLKSE-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2278 3 / PGII
WGK Germany 
2
RTECS 
MJ8840000
Flash Point(F) 
15.8 °F
Flash Point(C) 
-9 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?