EMAIL THIS PAGE TO A FRIEND
442274 Supelco

1-Octene

analytical standard

  • CAS Number 111-66-0

  • Linear Formula CH3(CH2)5CH=CH2

  • Molecular Weight 112.21

  •  Beilstein Registry Number 1734497

  •  MDL number MFCD00009548

  •  PubChem Substance ID 24867773

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, Chromatography,
grade   analytical standard
vapor density   3.9 (vs air)
vapor pressure   36 mmHg ( 38 °C)
InChI Key   KWKAKUADMBZCLK-UHFFFAOYSA-N
form   neat
autoignition temp.   446 °F
feature   standard type Calibration
expl. lim.   3.9 %
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.408(lit.)
  n20/D 1.409(lit.)
bp   122-123 °C(lit.)
mp   −101 °C (lit.)
density   0.715 g/mL at 25 °C(lit.)
Featured Industry   Cosmetics, Personal Care, and Cleaning Products
Food and Beverages
Petroleum
format   neat
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Repeated exposure may cause skin dryness or cracking.
RIDADR 
UN 3295BF 3 / PGII
WGK Germany 
3
Flash Point(F) 
69.8 °F
Flash Point(C) 
21 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?