• USA Home
  • 442291-U - 2,3-Dichlorophenol

EMAIL THIS PAGE TO A FRIEND
442291-U Supelco

2,3-Dichlorophenol

analytical standard

Purchase

Properties

grade   analytical standard
form   neat
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
bp   206 °C (lit.)
mp   55-57 °C (lit.)
Featured Industry   Agriculture
Environmental
format   neat
SMILES string   Oc1cccc(Cl)c1Cl
InChI   1S/C6H4Cl2O/c7-4-2-1-3-5(9)6(4)8/h1-3,9H
InChI key   UMPSXRYVXUPCOS-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 2020 6.1 / PGIII
WGK Germany 
3
RTECS 
SK8450000
Flash Point(F) 
239 °F
Flash Point(C) 
115 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Phenolic Calibration Standards

Phenolic compounds are important raw materials for the manufacturing of phenolic resins, pharmaceuticals, laboratory reagents, dyes, germicidal paints, and general disinfectants. Due to their toxicit...
Reporter US Volume 28.3
Keywords: Chromatography, Environmental, Gas chromatography, PAGE, Pharmaceutical, Sample preparations

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?