EMAIL THIS PAGE TO A FRIEND
442322 Supelco

2,5-Dimethylphenol

analytical standard

Synonym: 2-Hydroxy-p-xylene, p-Xylenol

  • CAS Number 95-87-4

  • Linear Formula (CH3)2C6H3OH

  • Molecular Weight 122.16

  •  Beilstein/REAXYS Number 1099260

  •  EC Number 202-461-5

  •  MDL number MFCD00002237

  •  eCl@ss 39023308

  •  PubChem Substance ID 24867798

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, Chromatography,
grade   analytical standard
form   neat
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
bp   212 °C (lit.)
Featured Industry   Agriculture
Environmental
format   neat
storage temp.   room temp
SMILES string   Cc1ccc(C)c(O)c1
InChI   1S/C8H10O/c1-6-3-4-7(2)8(9)5-6/h3-5,9H,1-2H3
InChI key   NKTOLZVEWDHZMU-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2261 6.1 / PGII
WGK Germany 
3
RTECS 
ZE5775000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?