EMAIL THIS PAGE TO A FRIEND
442342 Supelco

2-Chloroaniline

analytical standard

  • CAS Number 95-51-2

  • Linear Formula ClC6H4NH2

  • Molecular Weight 127.57

  •  Beilstein/REAXYS Number 606077

  •  EC Number 202-426-4

  •  MDL number MFCD00007656

  •  PubChem Substance ID 24867807

Purchase

Properties

grade   analytical standard
InChI Key   AKCRQHGQIJBRMN-UHFFFAOYSA-N
packaging   ampule of 1000 mg
bp   208-210 °C (lit.)
density   1.213 g/mL at 25 °C (lit.)
Featured Industry   Environmental
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2019 6.1 / PGII
WGK Germany 
3
RTECS 
BX0525000
Flash Point(F) 
208.4 °F
Flash Point(C) 
98 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?