EMAIL THIS PAGE TO A FRIEND
442359 Supelco

2-Methylnaphthalene

analytical standard

Synonym: β-Methylnaphthalene

  • CAS Number 91-57-6

  • Empirical Formula (Hill Notation) C11H10

  • Molecular Weight 142.20

  •  Beilstein/REAXYS Number 906859

  •  EC Number 202-078-3

  •  MDL number MFCD00004118

  •  PubChem Substance ID 24867811

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, Chemical Class,
grade   analytical standard
InChI Key   QIMMUPPBPVKWKM-UHFFFAOYSA-N
form   neat
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Environmental
format   neat
storage temp.   room temp
Gene Information   human ... CYP1A2(1544)

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2
RTECS 
QJ9635000
Flash Point(F) 
208.4 °F
Flash Point(C) 
98 °C

Documents

Certificate of Analysis

Please Enter a Lot Number
Protocols & Articles

Articles

GC Analysis of Aromatics in reforulated Gasoline on Equity®-1

From our library of Articles, Sigma-Aldrich presents GC Analysis of Aromatics in reforulated Gasoline on Equity®-1
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

GC Analysis of Aromatics in reforulated Gasoline on SLB®-IL111

From our library of Articles, Sigma-Aldrich presents GC Analysis of Aromatics in reforulated Gasoline on SLB®-IL111
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Separation

GC Analysis of Aromatics in reforulated Gasoline on SUPELCOWAX® 10

From our library of Articles, Sigma-Aldrich presents GC Analysis of Aromatics in reforulated Gasoline on SUPELCOWAX® 10
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

GC Analysis of Gasoline Residue in Arson Samples on a 100% Methyl Column after SPME using 100 μm PDMS Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Gasoline Residue in Arson Samples on a 100% Methyl Column after SPME using 100 μm PDMS Fiber
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Sample preparations, Size-exclusion chromatography, Solid phase microextractions

GC Analysis of Hydrocarbons in Gasoline on Equity®-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents GC Analysis of Hydrocarbons in Gasoline on Equity®-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Size-exclusion chromatography

GC Analysis of Hydrocarbons in Gasoline on Petrocol® DH, Isothermal

From our library of Articles, Sigma-Aldrich presents GC Analysis of Hydrocarbons in Gasoline on Petrocol® DH, Isothermal
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Separation

GC Analysis of Hydrocarbons in Gasoline on Petrocol® DH

From our library of Articles, Sigma-Aldrich presents GC Analysis of Hydrocarbons in Gasoline on Petrocol® DH
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Size-exclusion chromatography

GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) in Salmon on SPB®-608 (20 m x 0.18 mm I.D., 0.18 μm) after QuEChERS Cleanup using Supel™ QuE Z-Sep, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) in Salmon on SPB®-608 (20 m x 0.18 mm I.D., 0.18 μm) after QuEChERS Cleanup using Supel™ QuE Z-Sep, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Gas chromatography mass spectrometry, Mass spectrometry, Purification, Sample preparations, Solid phase extractions

US EPA Method 8270: GC Analysis of Semivolatiles on Equity®-5

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on Equity®-5
Keywords: Chromatography, Gas chromatography, Mass selective detector, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on SLB-5ms (20 m x 0.18 mm I.D., 0.36 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB-5ms (20 m x 0.18 mm I.D., 0.36 μm), Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification, Size-exclusion chromatography

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm), 28 °C/min Oven Ramp, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm), 28 °C/min Oven Ramp, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms, 350 °C Final Oven Temp.

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms, 350 °C Final Oven Temp.
Keywords: Chromatography, Gas chromatography

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method OLM04.2 SVOA: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method OLM04.2 SVOA: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?