• USA Home
  • 442407 - 4-Chloro-2-methylphenol

EMAIL THIS PAGE TO A FRIEND
442407 Supelco

4-Chloro-2-methylphenol

analytical standard

Synonym: 4-Chloro-o-cresol

Purchase

Properties

grade   analytical standard
form   neat
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
bp   220-225 °C (lit.)
mp   43-46 °C (lit.)
Featured Industry   Environmental
format   neat
storage temp.   room temp
SMILES string   Cc1cc(Cl)ccc1O
InChI   1S/C7H7ClO/c1-5-4-6(8)2-3-7(5)9/h2-4,9H,1H3
InChI key   RHPUJHQBPORFGV-UHFFFAOYSA-N
Gene Information   rat ... Ar(24208)

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN3437 - class 6.1 - PG 2 - EHS - Chlorocresols, solid
WGK Germany 
2
RTECS 
GO7120000
Flash Point(F) 
172.4 °F
Flash Point(C) 
78 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?