EMAIL THIS PAGE TO A FRIEND
442423 Supelco

5-Nitro-o-toluidine

analytical standard

Synonym: 2-Amino-4-nitrotoluene, 2-Methyl-5-nitroaniline, 5-Nitro-o-toluidine

  • CAS Number 99-55-8

  • Linear Formula CH3C6H3(NO2)NH2

  • Molecular Weight 152.15

  •  Beilstein Registry Number 879021

  •  EC Number 202-765-8

  •  MDL number MFCD00007741

  •  PubChem Substance ID 24867844

Purchase

Properties

Related Categories 2002/61/EC, Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
InChI Key   DSBIJCMXAIKKKI-UHFFFAOYSA-N
form   neat
feature   standard type Calibration
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
mp   103-106 °C (lit.)
Featured Industry   Cosmetics, Personal Care, and Cleaning Products
Environmental
Food and Beverages
format   neat
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2660 6.1 / PGIII
WGK Germany 
3
Protocols & Articles

Articles

GC Analysis of a 32-Component Anilines Mix on SLB®-IL59

From our library of Articles, Sigma-Aldrich presents GC Analysis of a 32-Component Anilines Mix on SLB®-IL59
Keywords: Chromatography, Gas chromatography, Mass selective detector

US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on Equity®-5 (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on Equity®-5 (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Mass selective detector, Purification

US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?