EMAIL THIS PAGE TO A FRIEND
442493 Supelco

Bromacil

analytical standard

Synonym: 5-Bromo-3-sec.-butyl-6-methyluracil

  • CAS Number 314-40-9

  • Empirical Formula (Hill Notation) C9H13BrN2O2

  • Molecular Weight 261.12

  •  Beilstein/REAXYS Number 6804755

  •  EC Number 206-245-1

  •  MDL number MFCD00055364

  •  PubChem Substance ID 24867871

Purchase

Properties

grade   analytical standard
form   neat
packaging   ampule of 250 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
Environmental
format   neat
storage temp.   room temp
SMILES string   CCC(C)N1C(=O)NC(C)=C(Br)C1=O
InChI   1S/C9H13BrN2O2/c1-4-5(2)12-8(13)7(10)6(3)11-9(12)14/h5H,4H2,1-3H3,(H,11,14)
InChI key   CTSLUCNDVMMDHG-UHFFFAOYSA-N
Gene Information   human ... EPHX2(2053)
mouse ... Ephx2(13850)

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2
RTECS 
YQ9100000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

GC Analysis of 'Positive List' Chemicals (Mix 5) on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents GC Analysis of 'Positive List' Chemicals (Mix 5) on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Herbicides, Pesticides, Purification

GC Analysis of Nitrogen-Containing Herbicides in Water on a 5% Phenyl Column after SPME using 85 μm Polyacrylate Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Nitrogen-Containing Herbicides in Water on a 5% Phenyl Column after SPME using 85 μm Polyacrylate Fiber
Keywords: Chromatography, Gas chromatography, Mass spectrometry, Purification, Size-exclusion chromatography, Solid phase microextractions

US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?