EMAIL THIS PAGE TO A FRIEND
442549 Supelco

Dibenzofuran

analytical standard, ampule of 100 mg

Synonym: Diphenylene oxide

  • CAS Number 132-64-9

  • Empirical Formula (Hill Notation) C12H8O

  • Molecular Weight 168.19

  •  Beilstein/REAXYS Number 121100

  •  EC Number 205-071-3

  •  MDL number MFCD00004968

  •  PubChem Substance ID 24867901

Purchase

Properties

grade   analytical standard
packaging   ampule of 100 mg
bp   154-155 °C/20 mmHg (lit.)
mp   80-82 °C (lit.)
Featured Industry   Environmental
SMILES string   c1ccc2c(c1)oc3ccccc23
InChI   1S/C12H8O/c1-3-7-11-9(5-1)10-6-2-4-8-12(10)13-11/h1-8H
InChI key   TXCDCPKCNAJMEE-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
HP4430000
Flash Point(F) 
266 °F
Flash Point(C) 
130 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?