• USA Home
  • 442560 - Dichloroacetonitrile

EMAIL THIS PAGE TO A FRIEND
442560 Supelco

Dichloroacetonitrile

analytical standard

Purchase

Properties

grade   analytical standard
form   neat
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.44 (lit.)
bp   110-112 °C (lit.)
density   1.369 g/mL at 25 °C (lit.)
Featured Industry   Environmental
format   neat
storage temp.   room temp
SMILES string   ClC(Cl)C#N
InChI   1S/C2HCl2N/c3-2(4)1-5/h2H
InChI key   STZZWJCGRKXEFF-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2920 3(8) / PGII
WGK Germany 
3
RTECS 
AL8465000
Flash Point(F) 
96.8 °F
Flash Point(C) 
36 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?