EMAIL THIS PAGE TO A FRIEND
442564 Supelco

Dicyclohexylamine

analytical standard

Purchase

Properties

Related Categories Additional Standards, Aliphatics, Alpha Sort, Amines, Analytical Standards,
grade   analytical standard
vapor density   6 (vs air)
vapor pressure   12 mmHg ( 37.7 °C)
InChI Key   XBPCUCUWBYBCDP-UHFFFAOYSA-N
form   neat
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.484 (lit.)
bp   117-120 °C/10 mmHg
  256 °C (lit.)
mp   −2 °C (lit.)
density   0.912 g/mL at 20 °C (lit.)
Featured Industry   Environmental
format   neat
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2565 8 / PGIII
WGK Germany 
2
RTECS 
HY4025000
Flash Point(F) 
204.8 °F
Flash Point(C) 
96 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?