EMAIL THIS PAGE TO A FRIEND
442570 Supelco

Dinoseb

PESTANAL®, analytical standard

Synonym: 2-sec-Butyl-4,6-dinitrophenol

Purchase

Properties

Related Categories A-D, Additional Pesticide Standards, Additional Standards, All Herbicide Standards, Alpha Index of All Pesticide Standards,
grade   analytical standard
InChI Key   OWZPCEFYPSAJFR-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
packaging   ampule of 100 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
Cleaning Products
Cosmetics
Food and Beverages
Personal Care
format   neat
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

Signal word 
Danger
Supplemental Hazard Statements 
Risk of explosion if heated under confinement.
RIDADR 
UN2811 - class 6.1 - PG 2 - MP - RQ - Toxic solids, organic, n.o.s. HI: all
WGK Germany 
3
RTECS 
SJ9800000
Flash Point(F) 
305.6 °F
Flash Point(C) 
152 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

GC Analysis of Phenols in Water on a 5% Phenyl Column after SPME using 85 μm Polyacrylate Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Phenols in Water on a 5% Phenyl Column after SPME using 85 μm Polyacrylate Fiber
Keywords: Chromatography, Gas chromatography, Mass spectrometry, Purification, Size-exclusion chromatography, Solid phase microextractions

HPLC Analysis of Acidic Herbicides in Water on a Polymeric C18 Column after SPE using Supelclean™ ENVI™-Carb

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Acidic Herbicides in Water on a Polymeric C18 Column after SPE using Supelclean™ ENVI™-Carb
Keywords: Chromatography, Environmental, Environmental Analysis, Herbicides, High performance liquid chromatography, Sample preparations, Size-exclusion chromatography, Solid phase extractions

US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on Equity®-5 (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on Equity®-5 (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Mass selective detector, Purification

US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?