EMAIL THIS PAGE TO A FRIEND
442598 Supelco

γ-BHC

analytical standard

Synonym: γ-BHC, 1α,2α,3β,4α,5α,6β-Hexachlorocyclohexane, Lindane

  • CAS Number 58-89-9

  • Empirical Formula (Hill Notation) C6H6Cl6

  • Molecular Weight 290.83

  •  Beilstein/REAXYS Number 1907337

  •  EC Number 200-401-2

  •  MDL number MFCD00135947

  •  PubChem Substance ID 24867924

Purchase

Properties

grade   analytical standard
form   neat
packaging   ampule of 250 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
mp   113-115 °C (lit.)
Featured Industry   Agriculture
Environmental
format   neat
SMILES string   Cl[C@H]1[C@H](Cl)[C@H](Cl)[C@@H](Cl)[C@H](Cl)[C@@H]1Cl
InChI   1S/C6H6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-6H/t1-,2-,3-,4+,5+,6+
InChI key   JLYXXMFPNIAWKQ-GNIYUCBRSA-N
Gene Information   mouse ... Esr1(13982)
rat ... Ar(24208)

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN2811 - class 6.1 - PG 3 - SMP - RQ - Toxic solid, organic, n.o.s., HI: all
WGK Germany 
3
RTECS 
GV4900000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?