• USA Home
  • 442656 - Methyl 2,3-dichloropropionate

EMAIL THIS PAGE TO A FRIEND
442656 Supelco

Methyl 2,3-dichloropropionate

analytical standard

  • CAS Number 3674-09-7

  • Linear Formula ClCH2CHClCOOCH3

  • Molecular Weight 157.00

  •  Beilstein Registry Number 1701891

  •  EC Number 222-940-2

  •  MDL number MFCD00000944

  •  PubChem Substance ID 24867941

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, Chromatography,
grade   analytical standard
InChI Key   OFHMODDLBXETIK-UHFFFAOYSA-N
form   neat
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.453(lit.)
bp   52-55 °C/2 mmHg(lit.)
density   1.325 g/mL at 25 °C(lit.)
Featured Industry   Environmental
format   neat
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
145.4 °F
Flash Point(C) 
63 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?