EMAIL THIS PAGE TO A FRIEND
442709 Supelco

Tetratetracontane

analytical standard

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, Chemical Class,
grade   analytical standard
description   This product has been discontinued. It has been replaced by product 88144.
InChI Key   KMXFZRSJMDYPPG-UHFFFAOYSA-N
form   neat
packaging   ampule of 500 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
mp   85-87 °C (lit.)
Featured Industry   Petroleum
format   neat
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

GC Analysis of Total Petroleum Hydrocarbons (TPH) on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents GC Analysis of Total Petroleum Hydrocarbons (TPH) on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

GC Analysis of Total Petroleum Hydrocarbons (TPH) on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents GC Analysis of Total Petroleum Hydrocarbons (TPH) on SLB®-5ms
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?