EMAIL THIS PAGE TO A FRIEND
442713 Supelco

Tridecane

analytical standard

  • CAS Number 629-50-5

  • Linear Formula CH3(CH2)11CH3

  • Molecular Weight 184.36

  •  Beilstein/REAXYS Number 1733089

  •  EC Number 211-093-4

  •  MDL number MFCD00008979

  •  PubChem Substance ID 24867979

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, Chemical Class,
grade   analytical standard
vapor density   6.4 (vs air)
vapor pressure   1 mmHg ( 59.4 °C)
InChI Key   IIYFAKIEWZDVMP-UHFFFAOYSA-N
form   neat
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.425 (lit.)
bp   110-112 °C/12 mmHg(lit.)
  234 °C(lit.)
mp   −6-−4 °C (lit.)
density   0.756 g/mL at 25 °C (lit.)
Featured Industry   Petroleum
format   neat
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Product 442713 has been discontinued. Product 91490 may be an acceptable replacement.

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Repeated exposure may cause skin dryness or cracking.
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
174.2 °F
Flash Point(C) 
79 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

GC Analysis of Aromatics in reforulated Gasoline on SLB®-IL111

From our library of Articles, Sigma-Aldrich presents GC Analysis of Aromatics in reforulated Gasoline on SLB®-IL111
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Separation

GC Analysis of Aromatics in reforulated Gasoline on SUPELCOWAX® 10

From our library of Articles, Sigma-Aldrich presents GC Analysis of Aromatics in reforulated Gasoline on SUPELCOWAX® 10
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

GC Analysis of BTEX, Undecane, and Tridecane on SLB®-IL100

From our library of Articles, Sigma-Aldrich presents GC Analysis of BTEX, Undecane, and Tridecane on SLB®-IL100
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

GC Analysis of Refinery Grade Unleaded Gasoline on TCEP (15 m x 0.10 mm I.D., 0.18 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents GC Analysis of Refinery Grade Unleaded Gasoline on TCEP (15 m x 0.10 mm I.D., 0.18 μm), Fast GC Analysis
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

GC Analysis of an Indoor Air Standard on Equity®-1

From our library of Articles, Sigma-Aldrich presents GC Analysis of an Indoor Air Standard on Equity®-1
Keywords: Air Monitoring, Chromatography, Gas chromatography, Purification, Size-exclusion chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?