EMAIL THIS PAGE TO A FRIEND
442714 Supelco

Undecane

analytical standard

Synonym: n-Undecane, Hendecane

  • CAS Number 1120-21-4

  • Linear Formula CH3(CH2)9CH3

  • Molecular Weight 156.31

  •  Beilstein Registry Number 1697099

  •  EC Number 214-300-6

  •  MDL number MFCD00008959

  •  PubChem Substance ID 24867980

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, Chemical Class,
grade   analytical standard
vapor density   5.4 (vs air)
vapor pressure   <0.4 mmHg ( 20 °C)
InChI Key   RSJKGSCJYJTIGS-UHFFFAOYSA-N
form   neat
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.417(lit.)
bp   196 °C(lit.)
mp   −26 °C (lit.)
density   0.74 g/mL at 25 °C(lit.)
Featured Industry   Petroleum
format   neat
storage temp.   room temp

Description

Other Notes

Product 442714 has been discontinued. Product 94000 may be an acceptable replacement.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Repeated exposure may cause skin dryness or cracking.
RIDADR 
UN 2330 3 / PGIII
WGK Germany 
3
RTECS 
YQ1525000
Flash Point(F) 
143.6 °F
Flash Point(C) 
62 °C
Protocols & Articles

Articles

GC Analysis of Aromatics in reforulated Gasoline on Equity®-1

From our library of Articles, Sigma-Aldrich presents GC Analysis of Aromatics in reforulated Gasoline on Equity®-1
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

GC Analysis of Aromatics in reforulated Gasoline on SUPELCOWAX® 10

From our library of Articles, Sigma-Aldrich presents GC Analysis of Aromatics in reforulated Gasoline on SUPELCOWAX® 10
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

GC Analysis of BTEX, Undecane, and Tridecane on SLB®-IL100

From our library of Articles, Sigma-Aldrich presents GC Analysis of BTEX, Undecane, and Tridecane on SLB®-IL100
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

GC Analysis of Semivolatiles on SLB®-5ms after SPME using 60 μm Carbowax® Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Semivolatiles on SLB®-5ms after SPME using 60 μm Carbowax® Fiber
Keywords: Chromatography, Gas chromatography, Purification, Solid phase microextractions

GC Analysis of Total Petroleum Hydrocarbons (TPH) on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents GC Analysis of Total Petroleum Hydrocarbons (TPH) on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

GC Analysis of Total Petroleum Hydrocarbons (TPH) on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents GC Analysis of Total Petroleum Hydrocarbons (TPH) on SLB®-5ms
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

GC Analysis of an Indoor Air Standard on Equity®-1

From our library of Articles, Sigma-Aldrich presents GC Analysis of an Indoor Air Standard on Equity®-1
Keywords: Air Monitoring, Chromatography, Gas chromatography, Purification, Size-exclusion chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?