EMAIL THIS PAGE TO A FRIEND
442723 Supelco

o-Toluidine

analytical standard

Synonym: 2-Aminotoluene, 2-Methylaniline

  • CAS Number 95-53-4

  • Linear Formula CH3C6H4NH2

  • Molecular Weight 107.15

  •  Beilstein/REAXYS Number 741981

  •  EC Number 202-429-0

  •  MDL number MFCD00007730

  •  PubChem Substance ID 24867986

Purchase

Properties

grade   analytical standard
vapor density   3.7 (vs air)
vapor pressure   0.26 mmHg ( 25 °C)
autoignition temp.   899 °F
packaging   ampule of 100 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.572 (lit.)
bp   199-200 °C (lit.)
  89-90 °C/11 mmHg (lit.)
mp   −28 °C (lit.)
density   1.008 g/mL at 25 °C (lit.)
Featured Industry   Environmental
SMILES string   Cc1ccccc1N
InChI   1S/C7H9N/c1-6-4-2-3-5-7(6)8/h2-5H,8H2,1H3
InChI key   RNVCVTLRINQCPJ-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1708 6.1 / PGII
WGK Germany 
3
RTECS 
XU2975000
Flash Point(F) 
185 °F
Flash Point(C) 
85 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?