EMAIL THIS PAGE TO A FRIEND
442730 Supelco

Oxamyl

analytical standard

  • CAS Number 23135-22-0

  • Empirical Formula (Hill Notation) C7H13N3O3S

  • Molecular Weight 219.26

  •  Beilstein Registry Number 2212753

  •  EC Number 245-445-3

  •  MDL number MFCD00055334

  •  PubChem Substance ID 24867989

Purchase

Properties

Related Categories Additional Standards, Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, Carbamates,
grade   analytical standard
InChI Key   KZAUOCCYDRDERY-UITAMQMPSA-N
form   neat
packaging   ampule of 100 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
Environmental
format   neat
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN2811 - class 6.1 - PG 1 - PIH - MP - RQ - Toxic solids, organic, n.o.s. HI: all
WGK Germany 
3
RTECS 
RP2300000

Documents

Certificate of Analysis

Protocols & Articles

Articles

HPLC Analysis of Carbamate and Urea Pesticides on SUPELCOSIL™ LC-8

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Carbamate and Urea Pesticides on SUPELCOSIL™ LC-8
Keywords: Chromatography, Environmental, Environmental Analysis, Herbicides, High performance liquid chromatography, Pesticides

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?