• USA Home
  • 442735 - p-Tolualdehyde-2,4-DNPH

EMAIL THIS PAGE TO A FRIEND
442735 Supelco

p-Tolualdehyde-2,4-DNPH

(aldehyde, equivalent), analytical standard

Synonym: p-Tolualdehyde 2,4-dinitrophenylhydrazone

Purchase

Properties

grade   analytical standard
InChI Key   KOMHUUXZDFABJZ-OQLLNIDSSA-N
form   neat
feature   standard type calibration
packaging   ampule of 100 mg
concentration   (aldehyde, equivalent)
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
mp   239-240 °C (lit.)
Featured Industry   Environmental
format   neat
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

General description

p-Tolualdehyde-2,4-DNPH may be used as a reference standard in the separation and analysis of p-tolualdehyde-2,4-DNPH from essential oils using high-performance liquid chromatography (HPLC).

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?