• USA Home
  • 442739-U - Pentachlorobenzene

EMAIL THIS PAGE TO A FRIEND
442739-U Supelco

Pentachlorobenzene

analytical standard

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, M-P, Organic Pollutant Standards, Volatile & Semivolatile Standards More...
grade   analytical standard
InChI Key   CEOCDNVZRAIOQZ-UHFFFAOYSA-N
form   neat
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
bp   275-277 °C(lit.)
mp   84-87 °C (lit.)
density   1.609 g/mL at 25 °C (lit.)
Featured Industry   Environmental
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1325 - class 4.1 - PG 2 - RQ - Flammable solids, organic, n.o.s., HI: all
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on Equity®-5 (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on Equity®-5 (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Mass selective detector, Purification

US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method TO-10/IP-8 and ASTM® D4861: GC Analysis of Pesticides on SPB®-608 after Collection/Desorption using ORBO™-1000

From our library of Articles, Sigma-Aldrich presents US EPA Method TO-10/IP-8 and ASTM® D4861: GC Analysis of Pesticides on SPB®-608 after Collection/Desorption using ORBO™-1000
Keywords: Air Monitoring, Chromatography, Electron capture detector, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?