EMAIL THIS PAGE TO A FRIEND
442744 Supelco

Pentafluorobenzene

analytical standard

Purchase

Properties

grade   analytical standard
InChI Key   WACNXHCZHTVBJM-UHFFFAOYSA-N
form   neat
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.391 (lit.)
bp   85 °C (lit.)
mp   −48 °C (lit.)
density   1.514 g/mL at 25 °C (lit.)
Featured Industry   Environmental
format   neat
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1993BF 3 / PGII
WGK Germany 
2
RTECS 
DA6651600
Flash Point(F) 
57.2 °F
Flash Point(C) 
14 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?