EMAIL THIS PAGE TO A FRIEND
442745 Supelco

Pentafluorophenol

analytical standard

Synonym: 2,3,4,5,6-Pentafluorophenol

  • CAS Number 771-61-9

  • Empirical Formula (Hill Notation) C6HF5O

  • Molecular Weight 184.06

  •  Beilstein/REAXYS Number 1912584

  •  EC Number 212-235-8

  •  MDL number MFCD00002156

  •  PubChem Substance ID 24867998

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, Chromatography,
grade   analytical standard
form   neat
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Environmental
format   neat
storage temp.   room temp
SMILES string   Oc1c(F)c(F)c(F)c(F)c1F
InChI   1S/C6HF5O/c7-1-2(8)4(10)6(12)5(11)3(1)9/h12H
InChI key   XBNGYFFABRKICK-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
SM6680000
Flash Point(F) 
161.6 °F
Flash Point(C) 
72 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Phenolic Calibration Standards

Phenolic compounds are important raw materials for the manufacturing of phenolic resins, pharmaceuticals, laboratory reagents, dyes, germicidal paints, and general disinfectants. Due to their toxicit...
Reporter US Volume 28.3
Keywords: Chromatography, Environmental, Gas chromatography, PAGE, Pharmaceutical, Sample preparations

US EPA Method 8041: GC Analysis of Phenols on Equity®-5

From our library of Articles, Sigma-Aldrich presents US EPA Method 8041: GC Analysis of Phenols on Equity®-5
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Size-exclusion chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?