EMAIL THIS PAGE TO A FRIEND
442746 Supelco

Pentane

analytical standard

  • CAS Number 109-66-0

  • Linear Formula CH3(CH2)3CH3

  • Molecular Weight 72.15

  •  Beilstein/REAXYS Number 969132

  •  EC Number 203-692-4

  •  MDL number MFCD00009498

  •  PubChem Substance ID 24867999

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, Chromatography,
Quality Level   100
grade   analytical standard
vapor density   2.48 (vs air)
description   This product has been discontinued. It has been replaced by product 76870.
form   neat
autoignition temp.   500 °F
expl. lim.   ~8.3 %
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.358 (lit.)
bp   35-36 °C (lit.)
mp   −130 °C (lit.)
density   0.626 g/mL at 25 °C (lit.)
Featured Industry   Environmental
Petroleum
format   neat
storage temp.   room temp
SMILES string   CCCCC
InChI   1S/C5H12/c1-3-5-4-2/h3-5H2,1-2H3
InChI key   OFBQJSOFQDEBGM-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Repeated exposure may cause skin dryness or cracking.
RIDADR 
UN1265 - class 3 - PG 2 - Pentanes
WGK Germany 
2
RTECS 
RZ9450000
Flash Point(F) 
-56.2 °F
Flash Point(C) 
-49 °C
Protocols & Articles

Articles

ASTM D5134: GC Analysis of Hydrocarbons on Petrocol® DH 50.2

This application demonstrates the suitability of the Petrocol column for the efficient analysis of hydrocarbons as listed in ASTM method D5134.
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Size-exclusion chromatography

GC Analysis of Aromatics in reforulated Gasoline on Equity®-1

From our library of Articles, Sigma-Aldrich presents GC Analysis of Aromatics in reforulated Gasoline on Equity®-1
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

GC Analysis of Class 3 Residual Solvents on Equity®-1 after SPME using 100 μm PDMS Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Class 3 Residual Solvents on Equity®-1 after SPME using 100 μm PDMS Fiber
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Pharmaceutical, Size-exclusion chromatography, Solid phase microextractions

GC Analysis of Class 3 Residual Solvents on Equity®-5

From our library of Articles, Sigma-Aldrich presents GC Analysis of Class 3 Residual Solvents on Equity®-5
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Pharmaceutical, Size-exclusion chromatography

GC Analysis of Class 3 Residual Solvents on OVI-G43

From our library of Articles, Sigma-Aldrich presents GC Analysis of Class 3 Residual Solvents on OVI-G43
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Pharmaceutical, Size-exclusion chromatography

GC Analysis of Class 3 Residual Solvents on SUPELCOWAX® 10

From our library of Articles, Sigma-Aldrich presents GC Analysis of Class 3 Residual Solvents on SUPELCOWAX® 10
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Pharmaceutical, Size-exclusion chromatography

GC Analysis of Class 3 Residual Solvents on VOCOL® after SPME using 100 μm PDMS Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Class 3 Residual Solvents on VOCOL® after SPME using 100 μm PDMS Fiber
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Pharmaceutical, Size-exclusion chromatography, Solid phase microextractions

GC Analysis of Hydrocarbons in Gasoline on Equity®-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents GC Analysis of Hydrocarbons in Gasoline on Equity®-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Size-exclusion chromatography

GC Analysis of Hydrocarbons in Gasoline on Petrocol® DH, Isothermal

From our library of Articles, Sigma-Aldrich presents GC Analysis of Hydrocarbons in Gasoline on Petrocol® DH, Isothermal
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Separation

GC Analysis of Hydrocarbons in Gasoline on Petrocol® DH

From our library of Articles, Sigma-Aldrich presents GC Analysis of Hydrocarbons in Gasoline on Petrocol® DH
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Size-exclusion chromatography

GC Analysis of Impurities in MTBE on Petrocol® DH

From our library of Articles, Sigma-Aldrich presents GC Analysis of Impurities in MTBE on Petrocol® DH
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Size-exclusion chromatography

US EPA Method 8015 (modified): GC Analysis of Gasoline Range Organics (GRO) on Equity®-1 after Purge & Trap using "M" Trap

From our library of Articles, Sigma-Aldrich presents US EPA Method 8015 (modified): GC Analysis of Gasoline Range Organics (GRO) on Equity®-1 after Purge & Trap using "M" Trap
Keywords: Chromatography, Environmental, Flame ionization detector, Gas chromatography, Purification, Solid phase microextractions

US EPA Method 8015 (modified): GC Analysis of Gasoline Range Organics (GRO) on VOCOL® after Purge & Trap using "M" Trap

From our library of Articles, Sigma-Aldrich presents US EPA Method 8015 (modified): GC Analysis of Gasoline Range Organics (GRO) on VOCOL® after Purge & Trap using "M" Trap
Keywords: Chromatography, Environmental, Flame ionization detector, Gas chromatography, Purification, Solid phase microextractions

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?