• USA Home
  • 442771-U - Prowl® (Pendimethaline)

EMAIL THIS PAGE TO A FRIEND
442771-U Supelco

Prowl® (Pendimethaline)

analytical standard

Synonym: Pendimethalin

  • CAS Number 40487-42-1

  • Empirical Formula (Hill Notation) C13H19N3O4

  • Molecular Weight 281.31

  •  Beilstein/REAXYS Number 2157711

  •  EC Number 254-938-2

  •  MDL number MFCD00055332

  •  PubChem Substance ID 329756487

Purchase

Properties

Related Categories All Herbicide Standards, Analytical Standards, Analytical/Chromatography, Dinitroaniline, Herbicides,
grade   analytical standard
InChI Key   CHIFOSRWCNZCFN-UHFFFAOYSA-N
form   neat
packaging   ampule of 250 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

Prowl is a registered trademark of BASF

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2
RTECS 
BX5470000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

GC Analysis of 'Positive List' Chemicals (Mix 3) on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents GC Analysis of 'Positive List' Chemicals (Mix 3) on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Herbicides, Pesticides, Purification

GC Analysis of Agricultural Pesticides on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents GC Analysis of Agricultural Pesticides on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Herbicides, Metabolites, Pesticides, Purification, Size-exclusion chromatography

GC Analysis of a 19-Component Pesticide Mix on SLB®-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents GC Analysis of a 19-Component Pesticide Mix on SLB®-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Pesticides, Purification

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?