EMAIL THIS PAGE TO A FRIEND
442802 Supelco

Thiophene

analytical standard

Synonym: Thiofuran, Thiole

  • CAS Number 110-02-1

  • Empirical Formula (Hill Notation) C4H4S

  • Molecular Weight 84.14

  •  Beilstein/REAXYS Number 103222

  •  EC Number 203-729-4

  •  MDL number MFCD00005413

  •  PubChem Substance ID 24867678

Purchase

Properties

grade   analytical standard
vapor density   2.9 (vs air)
vapor pressure   40 mmHg ( 12.5 °C)
InChI Key   YTPLMLYBLZKORZ-UHFFFAOYSA-N
form   neat
autoignition temp.   743 °F
expl. lim.   12.5 %
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.529 (lit.)
bp   84 °C(lit.)
mp   −38 °C (lit.)
density   1.051 g/mL at 25 °C (lit.)
Featured Industry   Environmental
format   neat
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2414 3 / PGII
WGK Germany 
3
RTECS 
XM7350000
Flash Point(F) 
30.2 °F
Flash Point(C) 
-1 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

GC Analysis of a 12-Component Sulfur Compound Mix on SLB®-IL59

From our library of Articles, Sigma-Aldrich presents GC Analysis of a 12-Component Sulfur Compound Mix on SLB®-IL59
Keywords: Chromatography, Gas chromatography, Mass selective detector, Size-exclusion chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?