• USA Home
  • 442821 - Trichloroacetonitrile

EMAIL THIS PAGE TO A FRIEND
442821 Supelco

Trichloroacetonitrile

analytical standard

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, Chemical Class,
grade   analytical standard
vapor pressure   58 mmHg ( 20 °C)
InChI Key   DRUIESSIVFYOMK-UHFFFAOYSA-N
form   neat
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.441(lit.)
bp   83-84 °C(lit.)
density   1.44 g/mL at 25 °C(lit.)
format   neat
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3276 6.1 / PGIII
WGK Germany 
3
RTECS 
AM2450000
Flash Point(F) 
383 °F
Flash Point(C) 
195 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?