• USA Home
  • 442833 - Tris(2,3-dibromopropyl) phosphate

EMAIL THIS PAGE TO A FRIEND
442833 Supelco

Tris(2,3-dibromopropyl) phosphate

analytical standard

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, Bromo,
grade   analytical standard
form   neat
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Environmental
format   neat
storage temp.   room temp
SMILES string   BrCC(Br)COP(=O)(OCC(Br)CBr)OCC(Br)CBr
InChI   1S/C9H15Br6O4P/c10-1-7(13)4-17-20(16,18-5-8(14)2-11)19-6-9(15)3-12/h7-9H,1-6H2
InChI key   PQYJRMFWJJONBO-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
3
RTECS 
UB0350000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Carcinogen Selector - Digestive System - Large Intestine

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Digestive System - Large Intestine
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Digestive System - Liver

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Digestive System - Liver
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Digestive System - Stomach

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Digestive System - Stomach
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Digestive System

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Digestive System
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Endocrine System - Liver Endocrine

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Endocrine System

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Reproductive System - Lung

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Respiratory System

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Urinary System - Kidney

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Urinary System

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?