• USA Home
  • 442870 - Triphenyltin chloride

EMAIL THIS PAGE TO A FRIEND
442870 Supelco

Triphenyltin chloride

analytical standard

Synonym: Chlorotriphenylstannane, Fentin chloride

  • CAS Number 639-58-7

  • Linear Formula (C6H5)3SnCl

  • Molecular Weight 385.47

  •  Beilstein Registry Number 524762

  •  EC Number 211-358-4

  •  MDL number MFCD00000519

  •  PubChem Substance ID 24868031

Purchase

Properties

grade   analytical standard
InChI Key   NJVOZLGKTAPUTQ-UHFFFAOYSA-M
packaging   ampule of 500 mg
bp   240 °C/13.5 mmHg(lit.)
mp   108 °C (dec.) (lit.)
Featured Industry   Agriculture
Environmental
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3146 6.1 / PGIII
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?