EMAIL THIS PAGE TO A FRIEND
44586-U Supelco

Isoparaffins Mix

(typical values shown), analytical standard

  •  NACRES NA.24

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Fuels and Hydrocarbons, PNA / PONA / P-I-A-N-O Standards, Petrochemical Standards More...
grade   analytical standard
packaging   ampule of 0.1 mL
concentration   (typical values shown)
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Agency/Method   ASTM® D6729
  ASTM® D6730
  ASTM® D6733
Featured Industry   Petroleum
format   multi-component solution
storage temp.   2-8°C

Description

General description

This is a 37 component mix of known isoparaffins. It is intended for use in identifying isoparaffins in complex hydrocarbon mixtures.

• Formulation is weight percent, and will vary lot to lot.
• A detailed data sheet listing components by weight percent, mole percent, and liquid volume percent, retention times and other formulaiton is provided with the product.
• Supplied in a crimp-top vial with hole cap and septa.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Analyte

Description

Product #

Add to Cart

3,3-Diethylpentane 1.57 wt. %    
2,3-Dimethylbutane .447 wt. %    
2,3-Dimethylheptane 1.482 wt. %    
2,5-Dimethylheptane 5.619 wt. %    
3,3-Dimethylheptane 1.677 wt. %    
3,4-Dimethylheptane (L) 1.964 wt. %    
3,5-Dimethylheptane (L) .0001 wt. %    
3,4-Dimethylheptane (D) 1.707 wt. %    
2,2-Dimethylhexane 1.313 wt. %    
2,3-Dimethylhexane 1.614 wt. %    
2,4-Dimethylhexane 1.646 wt. %    
2,5-Dimethylhexane 3.708 wt. %    
2,2-Dimethyloctane 3.253 wt. %    
2,3-Dimethyloctane 3.838 wt. %    
3,3-Dimethyloctane 3.174 wt. %    
2,2-Dimethylpentane 1.778 wt. %    
2,3-Dimethylpentane 1.781 wt. %    
2,4-Dimethylpentane 3.678 wt. %    
3,3-Dimethylpentane 1.86 wt. %    
3-Ethylhexane .711 wt. %    
3-Ethyloctane 3.687 wt. %    
3-Ethylpentane .526 wt. %    
2-Methylbutane 2.183 wt. %    
2-Methylheptane 4.367 wt. %    
3-Methylheptane 5.443 wt. %    
4-Methylheptane 3.198 wt. %    
2-Methylhexane 2.359 wt. %    
3-Methylhexane 1.602 wt. %    
2-Methylnonane 3.708 wt. %    
3-Methylnonane 5.756 wt. %    
2-Methyloctane 3.75 wt. %    
3-Methyloctane 5.598 wt. %    
2-Methylpentane 3.258 wt. %    
3-Methylpentane 5.356 wt. %    
2,2,3-Trimethylbutane 3.913 wt. %    
2,2,3-Trimethylpentane 1.721 wt. %    
Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
RIDADR 
UN 1993BE 3 / PGII
WGK Germany 
3
Flash Point(F) 
-9.4 °F
Flash Point(C) 
-23 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

analytical column

Product #

Description

Add to Cart

24160-U Petrocol® DH Capillary GC Column, L × I.D. 100 m × 0.25 mm, df 0.50 μm

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

standard

Product #

Description

Add to Cart

44594-U P-I-A-N-O Kit, analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?