EMAIL THIS PAGE TO A FRIEND
47236 Supelco

HMX solution

1000 μg/mL in acetonitrile, ampule of 1 mL

Synonym: 1,3,5,7-Tetranitro-1,3,5,7-tetrazocane solution, Cyclotetramethylene tetranitramine

  • CAS Number 2691-41-0

  • Empirical Formula (Hill Notation) C4H8N8O8

  • Molecular Weight 296.16

Purchase

Properties

feature   standard type calibration
packaging   ampule of 1 mL
concentration   1000 μg/mL in acetonitrile
Agency/Method   EPA 8330
storage temp.   2-8°C
Safety & Documentation

Safety Information

RIDADR 
UN 1648 3/PG 2
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?