• USA Home
  • 47241 - 3-Nitrotoluene solution

EMAIL THIS PAGE TO A FRIEND
47241 Supelco

3-Nitrotoluene solution

1000 μg/mL in acetonitrile, analytical standard

Purchase

Properties

Related Categories 8000 Series Solidwaste Methods, Additional Standards, Analytical Standards, Analytical/Chromatography, Aromatics,
grade   analytical standard
feature   standard type calibration
packaging   ampule of 1 mL
concentration   1000 μg/mL in acetonitrile
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Agency/Method   EPA 8330
Featured Industry   Environmental
format   single component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1648 3 / PGII
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles

Articles

EPA Method 8330: HPLC Analysis of Explosives on Ascentis® Express C18

From our library of Articles, Sigma-Aldrich presents EPA Method 8330: HPLC Analysis of Explosives on Ascentis® Express C18
Keywords: Chromatography, High performance liquid chromatography

GC Analysis of Explosives in Water on a 14% Cyanopropylphenyl Column after SPME using 65 μm PDMS/DVB Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Explosives in Water on a 14% Cyanopropylphenyl Column after SPME using 65 μm PDMS/DVB Fiber
Keywords: Chromatography, Electron capture detector, Gas chromatography, Purification, Solid phase microextractions

US EPA Method 8330: HPLC Analysis of Nitroaromatics and Nitroamines, Mix B on Ascentis® C18

From our library of Articles, Sigma-Aldrich presents US EPA Method 8330: HPLC Analysis of Nitroaromatics and Nitroamines, Mix B on Ascentis® C18
Keywords: Chromatography, Environmental, Environmental Analysis, High performance liquid chromatography

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?