• USA Home
  • 47274 - Volatile Organic Contaminants Mix 1

EMAIL THIS PAGE TO A FRIEND
47274 Supelco

Volatile Organic Contaminants Mix 1

certified reference material, 50 μg/mL each component in methanol

Purchase

Properties

Related Categories 500 Series Drinking Water Methods, Additional Standards, Analytical Standards, Analytical/Chromatography, Application Areas and Market Segments,
grade   certified reference material
  TraceCERT®
feature   standard type calibration
packaging   ampule of 1 mL
concentration   50 μg/mL each component in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Agency/Method   EPA 502.1
  EPA 503.1
  EPA 524.1
  EPA 524.2
Featured Industry   Environmental
format   multi-component solution
storage temp.   2-8°C

Description

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO Guide 34:2009 and ISO/IEC 17025:2005. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Analyte

Description

Product #

Add to Cart

Bromobenzene    
4-Chlorotoluene    
1,2-Dibromo-3-chloropropane    
1,2-Dibromoethane    
Dibromomethane    
2,2-Dichloropropane    
1,1-Dichloro-1-propene    
Styrene    
p-Xylene    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?