• USA Home
  • 47368-U - cis-1,3-Dichloropropene solution

EMAIL THIS PAGE TO A FRIEND
47368-U Supelco

cis-1,3-Dichloropropene solution

2000 μg/mL in methanol, analytical standard

  • CAS Number 10061-01-5

  • Empirical Formula (Hill Notation) C3H4Cl2

  • Molecular Weight 110.97

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, D, Organic Pollutant Standards, Volatile & Semivolatile Standards More...
grade   analytical standard
packaging   ampule of 1 mL
concentration   2000 μg/mL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Environmental
format   single component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
2
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

GC Analysis of Volatiles in Water on SPB®-1 after SPME using 100 μm PDMS Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Volatiles in Water on SPB®-1 after SPME using 100 μm PDMS Fiber
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Solid phase microextractions

GC Analysis of Volatiles on SPB®-624, Direct Injection

From our library of Articles, Sigma-Aldrich presents GC Analysis of Volatiles on SPB®-624, Direct Injection
Keywords: Chromatography, Gas chromatography, Purification, Size-exclusion chromatography

GC Analysis of Volatiles on VOCOL® after SPME using 75 μm Carboxen/PDMS Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Volatiles on VOCOL® after SPME using 75 μm Carboxen/PDMS Fiber
Keywords: Chromatography, Gas chromatography, Mass spectrometry, Purification, Size-exclusion chromatography, Solid phase microextractions

US EPA Method 502.2: GC Analysis of Volatiles on VOCOL® (105 m x 0.53 mm I.D.) after Purge & Trap using "K" Trap, ELCD Signal

From our library of Articles, Sigma-Aldrich presents US EPA Method 502.2: GC Analysis of Volatiles on VOCOL® (105 m x 0.53 mm I.D.) after Purge & Trap using "K" Trap, ELCD Signal
Keywords: Chromatography, Gas chromatography, Hall electrolytic conductivity detector, Photo-ionization detector, Purge and trap, Sample preparations

US EPA Method 502.2: GC Analysis of Volatiles on VOCOL® (105 m x 0.53 mm I.D.) after Purge & Trap using "K" Trap, PID Signal

From our library of Articles, Sigma-Aldrich presents US EPA Method 502.2: GC Analysis of Volatiles on VOCOL® (105 m x 0.53 mm I.D.) after Purge & Trap using "K" Trap, PID Signal
Keywords: Chromatography, Gas chromatography, Hall electrolytic conductivity detector, Photo-ionization detector, Purification

US EPA Method 524.2: GC Analysis of Volatiles on VOCOL® (105 m x 0.53 mm I.D.) after Purge & Trap using "K" Trap

From our library of Articles, Sigma-Aldrich presents US EPA Method 524.2: GC Analysis of Volatiles on VOCOL® (105 m x 0.53 mm I.D.) after Purge & Trap using "K" Trap
Keywords: Chromatography, Gas chromatography, Purification, Size-exclusion chromatography

US EPA Method 624: GC Analysis of Volatiles on SPB®-624 after Purge & Trap using "K" Trap, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 624: GC Analysis of Volatiles on SPB®-624 after Purge & Trap using "K" Trap, Fast GC Analysis
Keywords: Chromatography, Environmental, Gas chromatography, Purification, Solid phase microextractions

US EPA Method 624: GC Analysis of Volatiles on SPB®-624 after Purge & Trap using "K" Trap

From our library of Articles, Sigma-Aldrich presents US EPA Method 624: GC Analysis of Volatiles on SPB®-624 after Purge & Trap using "K" Trap
Keywords: Chromatography, Environmental, Gas chromatography, Purification, Solid phase microextractions

US EPA Method 624: GC Analysis of Volatiles on VOCOL® (60 m x 0.53 mm I.D.) after Purge & Trap using "K" Trap

From our library of Articles, Sigma-Aldrich presents US EPA Method 624: GC Analysis of Volatiles on VOCOL® (60 m x 0.53 mm I.D.) after Purge & Trap using "K" Trap
Keywords: Chromatography, Gas chromatography, Purification, Size-exclusion chromatography

US EPA Method 8260: GC Analysis of Volatiles on SPB®-624 after Purge & Trap using "K" Trap, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8260: GC Analysis of Volatiles on SPB®-624 after Purge & Trap using "K" Trap, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Gas chromatography mass spectrometry, Mass spectrometry, Purge and trap, Purification, Solid phase microextractions

US EPA Method 8260: GC Analysis of Volatiles on SPB®-624 after Purge & Trap using "K" Trap

From our library of Articles, Sigma-Aldrich presents US EPA Method 8260: GC Analysis of Volatiles on SPB®-624 after Purge & Trap using "K" Trap
Keywords: Chromatography, Environmental, Gas chromatography, Purification, Solid phase microextractions

US EPA Method 8260: GC Analysis of Volatiles on VOCOL® after Purge & Trap using "K" Trap, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8260: GC Analysis of Volatiles on VOCOL® after Purge & Trap using "K" Trap, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification, Solid phase microextractions

US EPA Method 8260: GC Analysis of Volatiles on VOCOL® after Purge & Trap using "K" Trap

From our library of Articles, Sigma-Aldrich presents US EPA Method 8260: GC Analysis of Volatiles on VOCOL® after Purge & Trap using "K" Trap
Keywords: Chromatography, Gas chromatography, Purification, Solid phase microextractions

US EPA Method OLM04.2 VOA: GC Analysis of Volatiles on SPB®-624 after Purge & Trap using "K" Trap, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method OLM04.2 VOA: GC Analysis of Volatiles on SPB®-624 after Purge & Trap using "K" Trap, Fast GC Analysis
Keywords: Chromatography, Environmental, Gas chromatography, Purification, Solid phase microextractions

US EPA Method OLM04.2 VOA: GC Analysis of Volatiles on VOCOL® after Purge & Trap using "K" Trap, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method OLM04.2 VOA: GC Analysis of Volatiles on VOCOL® after Purge & Trap using "K" Trap, Fast GC Analysis
Keywords: Chromatography, Environmental, Gas chromatography, Purification, Solid phase microextractions

US EPA Method TO-17: GC Analysis of Volatiles on Equity®-1 after Collection/Desorption using Carbotrap 300 Tube

From our library of Articles, Sigma-Aldrich presents US EPA Method TO-17: GC Analysis of Volatiles on Equity®-1 after Collection/Desorption using Carbotrap 300 Tube
Keywords: Chromatography, Gas chromatography

US EPA Method TO-17: GC Analysis of Volatiles on VOCOL® after Collection/Desorption using Air Toxics Tube

From our library of Articles, Sigma-Aldrich presents US EPA Method TO-17: GC Analysis of Volatiles on VOCOL® after Collection/Desorption using Air Toxics Tube
Keywords: Air Monitoring, Chromatography, Flame ionization detector, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?