• USA Home
  • 47431-U - EPA 601 Purgeable Mix w/o gases including 2-CEVE

EMAIL THIS PAGE TO A FRIEND
47431-U Supelco

EPA 601 Purgeable Mix w/o gases including 2-CEVE

certified reference material, 2000 μg/mL each component in methanol

  •  NACRES NA.24

Purchase

Properties

Related Categories 600 Series Wastewater Methods, Additional Standards, Analytical Standards, Analytical/Chromatography, Application Areas and Market Segments,
grade   certified reference material
description   without gases
grade   TraceCERT®
feature   standard type calibration
packaging   ampule of 1 mL
concentration   2000 μg/mL each component in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Agency/Method   EPA 601
Featured Industry   Environmental
format   multi-component solution
storage temp.   −20°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Analyte

Description

Product #

Add to Cart

Bromodichloromethane    
Bromoform    
Carbon tetrachloride    
Chlorobenzene    
2-Chloroethyl vinyl ether    
Chloroform    
Dibromochloromethane    
1,2-Dichlorobenzene    
1,3-Dichlorobenzene    
1,4-Dichlorobenzene    
1,1-Dichloroethane    
1,2-Dichloroethane    
1,1-Dichloroethylene    
trans-1,2-Dichloroethylene    
Dichloromethane    
1,2-Dichloropropane    
cis-1,3-Dichloropropene    
trans-1,3-Dichloropropene    
1,1,2,2-Tetrachloroethane    
Tetrachloroethylene    
1,1,1-Trichloroethane    
1,1,2-Trichloroethane    
Trichloroethylene    
Safety & Documentation

Safety Information

Signal word 
Danger
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
3
Flash Point(F) 
48.2 °F
Flash Point(C) 
9 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

06693 Timestrip Plus -20 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?