• USA Home
  • 47581-U - 2-Fluorobiphenyl solution

EMAIL THIS PAGE TO A FRIEND
47581-U Supelco

2-Fluorobiphenyl solution

certified reference material, 10 mg/mL in dichloromethane

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Certified Reference Materials (CRMs), Chemical Class,
grade   certified reference material
feature   standard type surrogate
packaging   ampule of 1 mL
concentration   10 mg/mL in dichloromethane
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Environmental
Petroleum
format   single component solution

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1593 6.1 / PGIII
WGK Germany 
2

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

US EPA Method 610: GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 610: GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) on SLB®-5ms
Keywords: Chromatography, Gas chromatography, High performance liquid chromatography, Purification, Separation

US EPA Method 610: GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) on SPB®-608

From our library of Articles, Sigma-Aldrich presents US EPA Method 610: GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) on SPB®-608
Keywords: Chromatography, Gas chromatography, High performance liquid chromatography, Purification, Separation

US EPA Method 625: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 625: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on Equity®-5 (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on Equity®-5 (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Mass selective detector, Purification

US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on Equity®-5

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on Equity®-5
Keywords: Chromatography, Gas chromatography, Mass selective detector, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on SLB-5ms (20 m x 0.18 mm I.D., 0.36 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB-5ms (20 m x 0.18 mm I.D., 0.36 μm), Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification, Size-exclusion chromatography

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm), 28 °C/min Oven Ramp, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm), 28 °C/min Oven Ramp, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms, 350 °C Final Oven Temp.

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms, 350 °C Final Oven Temp.
Keywords: Chromatography, Gas chromatography

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method OLM04.2 SVOA: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method OLM04.2 SVOA: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?