• USA Home
  • 47738-U - ASTM® D5580 Calibration Mix 4

EMAIL THIS PAGE TO A FRIEND
47738-U Supelco

ASTM® D5580 Calibration Mix 4

with Internal Standard, (Each component at nominal wt.% indicated.), analytical standard

  •  NACRES NA.24

Purchase

Properties

Related Categories ASTM D5580, ASTM Petroleum Standards, ASTM Petroleum and Petrochemical, Additional Standards, Analytical Standards,
grade   analytical standard
packaging   pkg of 1 × 9 mL
concentration   (Each component at nominal wt.% indicated.)
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Agency/Method   ASTM® D5580
Featured Industry   Petroleum
format   multi-component solution
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Analyte

Description

Product #

Add to Cart

Benzene 2 % (w/w)    
Ethylbenzene 5 % (w/w)    
Toluene 2.5 % (w/w)    
1,2,4-Trimethylbenzene 5 % (w/w)    
2,2,4-Trimethylpentane 80.5 % (w/w)    
o-Xylene 5 % (w/w)    
Safety & Documentation

Safety Information

Signal word 
Danger
Precautionary statements 
RIDADR 
UN1262 - class 3 - PG 2 - Octanes, solution
WGK Germany 
3
Flash Point(F) 
10.4 °F
Flash Point(C) 
-12 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?