• USA Home
  • 47761 - Methyl bromodichloroacetate solution

EMAIL THIS PAGE TO A FRIEND
47761 Supelco

Methyl bromodichloroacetate solution

1000 μg/mL in methyl tert-butyl ether, analytical standard

Purchase

Properties

Related Categories 500 Series Drinking Water Methods, Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
CofA   certificate of analysis is enclosed in each package.
feature   standard type calibration
packaging   ampule of 1 mL
concentration   1000 μg/mL in methyl tert-butyl ether
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Agency/Method   EPA 552.2
Featured Industry   Environmental
format   single component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
UN 2398 3 / PGII
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles

Articles

Drinking Water Calibration Standards for Monitoring Disinfectant Byproducts

However, disinfection byproducts are formed when chlorine and other chemical disinfectants react with naturally present organic compounds in the water. These byproducts have the potential to produce ...
Vicki Yearick
AnalytiX Volume 6 Article 1

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?