• USA Home
  • 47801 - Grain Fatty Acid Methyl Ester Mix

EMAIL THIS PAGE TO A FRIEND
47801 Supelco

Grain Fatty Acid Methyl Ester Mix

10 mg/mL in methylene chloride (as total weight), analytical standard

Purchase

Properties

grade   analytical standard
form   liquid
CofA   certificate of analysis is enclosed in each package.
packaging   pkg of 1 mL
concentration   10 mg/mL in methylene chloride (as total weight)
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   multi-component solution
functional group   ester
  methyl
storage temp.   −20°C

Description

General description

This fatty acid methyl ester (FAME) mixture is carefully prepared by weight. The weight percentage of each component is indicated. Each ampule contains 10 mg/mL of the FAME reference standard mix in methylene chloride.

For information on the chromatographic analysis of this standard, please contact Technical Services at techservice@sial.com.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools

Analyte

Description

Product #

Add to Cart

Methyl arachidate 1.9 wt. %    
Methyl behenate 1.9 wt. %    
Methyl caprylate 1.9 wt. %    
Methyl decanoate 3.2 wt. %    
Methyl cis-11-eicosenoate 1.9 wt. %    
Methyl elaidate 2.6 wt. %    
Methyl erucate 1.9 wt. %    
Methyl heptadecanoate 3.2 wt. %    
Methyl laurate 6.4 wt. %    
Methyl linoleate 13 wt. %    
Methyl linolenate 6.4 wt. %    
Methyl myristate 3.2 wt. %    
Methyl myristoleate 1.9 wt. %    
Methyl oleate 19.6 wt. %    
Methyl palmitate 13 wt. %    
Methyl palmitoleate 6.4 wt. %    
Methyl pentadecanoate 1.9 wt. %    
Methyl stearate 6.5 wt. %    
Methyl tridecanoate 3.2 wt. %    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Target organs 
Blood, Central nervous system, Liver
RIDADR 
UN 1593 6.1 / PGIII
WGK Germany 
2

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

CRM47801 Grain FAME Mix, certified reference material, in dichloromethane, ampule of 1 mL

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?